Learn More
Cayman Chemical ANTIBODY WZB117 25mg
SDP
Supplier:  Cayman Chemical 1990025 MG
Encompass_Preferred
An irreversible inhibitor of Glut1 that blocks glucose transport in diverse cancer cells (IC50 = ~0.6 µM) and cancer cell proliferation (IC50s= 5-10 µM); reduces tumor size in mice, with only small reductions in body weight and leukocyte count
Catalog No. 50-298-0926