Learn More
Cayman Chemical ANTIBODY X-Gal 5g
SDP
Supplier:  Cayman Chemical 164955 G
Encompass_Preferred
A chromogenic substrate for β-galactosidase; upon cleavage, a blue pigment is released that can be used as a marker of β-galactosidase activity; has been used for the detection of lacZ activity in reporter assays and in the determination of protein-protein interactions
Catalog No. 50-296-9294