Learn More
Cayman Chemical ANTIBODY X-nuNAc 1mg
SDP
Supplier:  Cayman Chemical 242061 MG
Encompass_Preferred
A chromogenic substrate for neuraminidases; the dark blue pigment 5,5’-dibromo-4-4’-dichloroindigo is released upon hydrolysis and can be used to quantify neuraminidase activity
Catalog No. 50-293-5871