Learn More
Cayman Chemical ANTIBODY XL019 25mg
SDP
Supplier:  Cayman Chemical 1629825 MG
Encompass_Preferred
A potent, bioavailable JAK inhibitor with preference for JAK2 (IC50 = 2 nM) over JAK1 and JAK3 (IC50s = 130 and 250 nM, respectively); effective against JAK2V617F as well as JAK2 and inhibits phosphorylation of STAT5 in vivo
Catalog No. 50-297-2626