Learn More
Cayman Chemical ANTIBODY XMD16-5 1mg
SDP
Supplier:  Cayman Chemical 233531 MG
Encompass_Preferred
A TNK2 inhibitor (IC50 = 0.38 µM); decreases viability of Ba/F3 cells expressing the oncogenic TNK2 D163E and R806Q mutant proteins (IC50s = 16 and 77 nM, respectively)
Catalog No. 50-299-3697