Learn More
Cayman Chemical ANTIBODY XMU-MP-1 1mg
SDP
Supplier:  Cayman Chemical 220831 MG
Encompass_Preferred
An inhibitor of MST1 and MST2 (IC50s = 71.1 and 38.1 nM, respectively); is selective for MST1 and 2 over a panel of 468 kinases at a concentration of 1 μM; inhibits phosphorylation of MOB1, LATS1 and 2, and YAP in HepG2 cells as well as MST1 and 2 autophosphorylation in RAW 264.7, U2OS, SW480, RPE1, SNU423, and HepG2 cells in a concentration-dependent manner; prevents cell death induced by overexpression of MST2; enhances liver regeneration in mice post partial hepatectomy and reduces liver fibrosis in a mouse model of chronic liver injury when administered at a dose of 1 mg/kg
Catalog No. 50-298-7590