Learn More
Cayman Chemical ANTIBODY XylazIn 100mg
SDP
Supplier:  Cayman Chemical 22641100 MG
Encompass_Preferred
An analytical reference standard categorized as an anesthetic; used as an adulterant in drugs of abuse and misused, leading to non-fatal and fatal intoxications; intended for use in analytical forensic applications
Catalog No. 50-299-5356