Learn More
Cayman Chemical ANTIBODY YH 239-EE 25mg
SDP
Supplier:  Cayman Chemical 2767825 MG
Encompass_Preferred
A prodrug form of YH 239; inhibits the growth of OCI-AML-3 cells and halts the cell cycle at the sub-G1 phase and induces apoptosis in NB4 and MOLM-13 cells at 20 μM
Catalog No. 50-293-8846