Learn More
Cayman Chemical ANTIBODY YM-155 10mg
SDP
Supplier:  Cayman Chemical 1149010 MG
Encompass_Preferred
A small molecule that suppresses transactivation of survivin through direct binding to its promoter; induces apoptosis in p53-deficient cancer cells in vitro at 10 nM with little effect on expression levels of other IAP- or Bcl-2-related proteins
Catalog No. 50-295-2101