Learn More
Cayman Chemical ANTIBODY YSK05 10mg
SDP
Supplier:  Cayman Chemical 3578610 MG
Encompass_Preferred
An ionizable cationic amino lipid (apparent pKa = ~6.5); has been used in combination with other lipids in the formation of liposomes and LNPs; administration of cyclic di-GMP in YSK05-containing liposomes increases serum levels of IFN-β, IFN-γ, IL-6, and TNF-α and activates NK cells in mice, as well as decreases lung metastasis in a B16/F10 murine melanoma model of metastasis; intravenous administration of factor VII siRNA in YSK05-containing LNPs reduces factor VII activity in isolated mouse plasma
Catalog No. 50-291-1214