Learn More
Cayman Chemical ANTIBODY YW1159 10mg
SDP
Supplier:  Cayman Chemical 2745110 MG
Encompass_Preferred
An inhibitor of Wnt signaling (IC50 = 1.2 nM); not cytotoxic to HEK293 cells following incubations of up to 24 hours but cell viability is decreased in a concentration-dependent manner after 72 hours
Catalog No. 50-293-8512