Learn More
Cayman Chemical ANTIBODY ZIdovudIn 100mg
SDP
Supplier:  Cayman Chemical 15492100 MG
Encompass_Preferred
An NRTI; reduces HIV-1 p24 antigen levels in the culture supernatant of HIV-1-infected isolated human peripheral blood monocytes at 0.01, 0.1, and 1 µM
Catalog No. 50-297-3050