Learn More
Cayman Chemical ANTIBODY ZInpyr-1 1mg
SDP
Supplier:  Cayman Chemical 151221 MG
Encompass_Preferred
A cell-permeable, fluorescein-based probe for zinc detection; undergoes a shift in excitation from 515 to 507 nm with increasing zinc concentrations using an emission filter over 513-558 nm
Catalog No. 50-297-2572