Learn More
Cayman Chemical ANTIBODY ZL006 50mg
SDP
Supplier:  Cayman Chemical 3414750 MG
Encompass_Preferred
An nNOS/PSD-95 interaction inhibitor; inhibits NMDA-induced lactate dehydrogenase release (IC50 = 0.082 µM), as well as glutamate and glycine-induced, NMDA receptor-dependent production of NO, in primary mouse neurons; prevents ischemia-induced increases in nNOS-PSD-95 complex formation in cortices isolated from a mouse model of MCAO-induced ischemia-reperfusion injury; reduces infarct size and increases survival in the same model; suppresses mechanical and cold allodynia in a mouse model of paclitaxel-induced neuropathic pain
Catalog No. 50-294-9476