Learn More
Cayman Chemical ANTIBODY ZSTK474 5mg
SDP
Supplier:  Cayman Chemical 173815 MG
Encompass_Preferred
A selective inhibitor of class I PI3K isoforms (IC50s = 17, 53, and 6 nM for p110β, -γ, and -δ, respectively); orally bioavailable, showing strong antitumor activity against human cancer xenografts in mice
Catalog No. 50-298-0612