Learn More
Cayman Chemical ANTIBODY11-HETE 25ug
SDP
Supplier:  Cayman Chemical 3450025 UG
Encompass_Preferred
A non-enzymatically derived oxylipin; levels are increased in an in vitro model of lipid peroxidation induced by FAS in rat brain homogenates and in rat liver in an in vivo model of lipid peroxidation induced by CCl4; has been found in skin extracts from individuals with psoriasis and in atherosclerotic plaques
Catalog No. 50-294-6296