Learn More
Cayman Chemical ANTIBODY12-HETE 50ug
SDP
Supplier:  Cayman Chemical 3455050 UG
Encompass_Preferred
A racemic mixture of arachidonic acid metabolites; induces aggregation of isolated human neutrophils at 1 µM; production of (±)12-HETE by platelets isolated from spontaneously hypertensive rats is increased compared with platelets isolated from normotensive rats
Catalog No. 50-294-6129