Learn More
Cayman Chemical ANTIBODY15-HETE 25ug
SDP
Supplier:  Cayman Chemical 3470025 UG
Encompass_Preferred
(±)15-HETE is one of the six monohydroxy fatty acids produced by the non-enzymatic oxidation of arachidonic acid. The biological activity of (±)15-HETE is similar to that of its constituent enantiomers (Item Nos. 34720 and 34710).
Catalog No. 50-291-0742