Learn More
Cayman Chemical ANTIBODY16-HDHA 25ug
SDP
Supplier:  Cayman Chemical 3360025 UG
Encompass_Preferred
(±)16-HDHA is an autoxidation product of DHA in vitro.{7479,7491} It is also produced from incubations of DHA in rat liver, brain, and intestinal microsomes.{7480,7490,7492} (±)16-HDHA is a potential marker of oxidative stress in brain and retina where DHA is an abundant PUFA.
Catalog No. 50-294-9885