Learn More
Cayman Chemical ANTIBODY18-HEPE 100ug
SDP
Supplier:  Cayman Chemical 32840100 UG
Encompass_Preferred
A non-enzymatic oxidation product of EPA; increased in the plasma and heart, as well as in the supernatant of cultured peritoneal macrophages isolated from fat-1 transgenic mice; reduces increases in cardiac fibrosis and the expression of Nppa, Col1a1, Tgfb1, Cx3cl1, and Adgre1 induced by transverse aortic constriction in a mouse model of cardiac hypertrophy at 1 and 5 UG
Catalog No. 50-294-7955