Learn More
Cayman Chemical ANTIBODY4-HDHA 25ug
SDP
Supplier:  Cayman Chemical 3320025 UG
Encompass_Preferred
An autoxidation product of DHA in vitro that can also be produced from incubation of DHA with rat liver, brain, and intestinal microsomes and enzymatic transformation of DHA by RBL-1 cells; is a potential marker of oxidative stress in brain and retina where DHA is an abundant PUFA
Catalog No. 50-295-0540