Learn More
Cayman Chemical ANTIBODY5 6-EET 25UG
SDP
Supplier:  Cayman Chemical 5021125 UG
Encompass_Preferred
A racemic version of a CYP450 metabolite of arachidonic acid; provided as a mixture of free acid and lactone; inhibits Cav3 channels (IC50 = 0.54 µM for Cav3.2); decreases nifedipine-resistant phenylephrine-induced vasoconstriction in isolated mouse mesenteric arteries via Cav3.2 blockade at 3 µM; a substrate of COX-1 and COX-2 in oxygen consumption and product formation assays at 50 µM
Catalog No. 50-289-3012