Learn More
Cayman Chemical ANTIBODY7-HDHA 100ug
SDP
Supplier:  Cayman Chemical 33300100 UG
Encompass_Preferred
(±)7-HDHA is an autoxidation product of DHA in vitro.{7479,7491} It is also produced from incubations of DHA in rat liver, brain, and intestinal microsomes.{7480,7490,7492} Enzymatic transformation of DHA by RBL-1 cells also produces 7-HDHA.{7515,7485} However, the enzymatic product is most likely the S-isomer. (±)7-HDHA is a potential marker of oxidative stress in brain and retina where DHA is an abundant PUFA.
Catalog No. 50-294-7234