Learn More
Cayman Chemical ANTIBODY9-HETE 250ug
SDP
Supplier:  Cayman Chemical 34400250 UG
Encompass_Preferred
A monohydroxy fatty acid; formed by lipid peroxidation of arachidonic acid, as well as by rat liver microsomal CYP; plasma levels are elevated in patients with angiographically defined coronary artery disease
Catalog No. 50-294-6872