Learn More
Cayman Chemical ANTIBODY9-HODE 1mg
SDP
Supplier:  Cayman Chemical 384001 MG
Encompass_Preferred
A acemic monohydroxy fatty acids resulting from the non-enzymatic oxidation of linoleic acid
Catalog No. 50-291-7192