Learn More
Cayman Chemical ANTIBODYS-DO271 5mg
SDP
Supplier:  Cayman Chemical 300385 MG
Encompass_Preferred
An inactive control for DO264; does not inhibit ABHD12-dependent hydrolysis of lyso-PS in mouse brain membrane lysates at 1 µM or modify inflammatory cytokine production in M1-polarized THP-1 macrophages
Catalog No. 50-299-0820