Learn More
Cayman Chemical ANTIBODYS-MG115 10mg
SDP
Supplier:  Cayman Chemical 1541310 MG
Encompass_Preferred
A potent and reversible proteasome inhibitor, targeting the chymotryptic site on the 20S particle (Ki = 21 nM); reduces the degradation of ubiquitin-conjugated proteins in extracts; blocks the degradation of long- and short-lived proteins in intact cells as well as the proteolytic generation of diverse proteins
Catalog No. 50-296-8923