Learn More
Medchemexpress LLC Antide | 112568-12-4 | 99.53% | 1591.29 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP0013
Antide (D-21074) is a potent LHRH antagonist. It can also be used for the research of prostatic cancer.
- Potent LHRH antagonist
- Used for prostatic cancer research
- Induces long-term chemical castration in adult male rats and cynomolgus monkeys
- Sealed storage, away from moisture and light
- For powder: -80°C for 2 years; -20°C for 1 year
- In solvent: -80°C for 6 months; -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.