Learn More
Sigma Aldrich Fine Chemicals Biosciences Antifoam 204, MFCD00130523, 50mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A831150ML
Encompass
Antifoam 204 is an organic antifoam used to prevent or minimize foaming in both microbial fermentation systems and mammalian suspension cultures. Control of foaming minimizes damage to the cells, resulting in increased protein/antibody production.
Catalog No. NC1685023