Learn More
Vector Laboratories Antigen Unmasking Solution, Citric Acid Based 250 ml
Supplier: Vector Laboratories H3300250
Antigen Unmasking Solutions are highly effective at revealing antigens in formalin-fixed, paraffin-embedded tissue sections when used in combination with a high temperature treatment procedure. This citrate-based solution (H-3300) is pH 6.0 and is supplied as convenient 100x concentrated stock, sufficient for preparation of 25 L of working solution. 250 ml