Learn More
American Research Products Inc AntiHuman IL2R alpha
Supplier:  American Research Products Inc IVMB049450UG
Encompass
Anti-Human IL-2R alpha (CD25) (Basiliximab) - APC Biosimilar is a biosimilar antibody with human/rhesus monkey/cynomolgus monkey reactivity Available in 50ug size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06004