Learn More
American Research Products Inc AntiHuman/Mouse CDCP1
Supplier:  American Research Products Inc IVMB03491MG
Encompass
Anti-Human/Mouse CDCP1 (CD318) In Vivo Antibody - Ultra Low Endotoxin is a in vivo antibody with human mouse reactivity produced in mouse Available in 1 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06110