Learn More
Enzo Life Sciences Antimycin A (5mg). CAS: 1397-94-0
SDP
Supplier:  Enzo Life Sciences ALX380075M005
Encompass_Preferred
Antibiotic. Induces apoptosis, which is not prevented by the presence of Bcl-2. Inhibits mitochondrial electron transport specifically between cytochromes b and c1. Alternative name: Antipiricullin, Virosin. Appearance: White to faint yellow powder. Solubility: Soluble in 100% ethanol (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8947