Learn More
Medchemexpress LLC HY-128553 5mg Medchemexpress, Antineoplaston A10 CAS:91531-30-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1285535MG
Encompass_Preferred
Medchemexpress, HY-128553 5mg Antineoplaston A10 CAS:91531-30-5 Antineoplaston A10, a naturally occurring substance in human body, is a Ras inhibitor potentially for the treatment of glioma, lymphoma, astrocytoma and breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9751