Learn More
Cayman Chemical Antioxidant Assay Kit, 96 wells
SDP
Supplier:  Cayman Chemical 70900196WELLS
Encompass_Preferred
Cayman’s Antioxidant Assay measures the total antioxidant capacity of plasma, serum, urine, saliva, or cell lysates. The assay relies on the ability of antioxidants in the sample to inhibit the oxidation of ABTS (2,2'-azino-di-[3-ethylbenzthiazoline sulphonate]) to ABTS•+ by metmyoglobin. The capacity of the antioxidants in the sample to prevent ABTS oxidation is compared with that of Trolox, a water-soluble tocopherol analogue, and is quantified as molar Trolox equivalents.
Catalog No. NC0692667