Learn More
Carbosynth LLC. Antipain, 100mg, MW: 604.70 • 36.46 • 36.03, Molecular formula: C27H44N10O6 • HCI • 2H2O
SDP
Supplier:  Carbosynth LLC. IAP4062100MG
Encompass_Preferred
Synonyms: [(S)-1-Carboxy-2-Phenylethyl]-Carbamoyl-L-Arginyl-L-Valyl-Argininal • Monohydrochloride Dihydrate, (S)-1-Carboxy-2-phenylethyl]-carbamoyl-Arg-Val-Arg-H • HCl • 2H,2O, Application: Microbial Product Inhibitor for Trypsin, u-PA, Papain, and Cathepsin A/B, Storage Temperature: -20°C, References: H. Suda, T. Aoyagi, M. Hamada, T. Takeuchi, and H. Umezawa, J. Antibiotics, 25, 263 (1972). (Original), S. Umezawa, K. Tatsuta, K. Fujimoto, T. Tsuchiya, H. Umezawa, and H. Naganawa, J. Antibiotics, 25, 267 (1972). (Original; Chem. Structure), J. Chau, J. Biol. Chem., 258, 4434 (1983). (Inhibitory Activity)
Catalog No. 50-168-5813