Learn More
Medchemexpress LLC HY-B0171 500mg Medchemexpress, Antipyrine CAS:60-80-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0171500MG
Encompass_Preferred
Medchemexpress, HY-B0171 500mg Antipyrine CAS:60-80-0 Antipyrine (Phenazone) is an antipyretic and analgesic. Antipyrine can be used as a probe drug for oxidative drug metabolism. Antipyrine has been widely used in assessment of hepatic oxidative capacity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.