Learn More
Med Vet International Antiskid Drill Bit, 1/16”, 1.5mm x 6cm, J0372AX, Each
Supplier:  Med Vet International JOR372AX
Encompass_Preferred
These drill bits have a sharp needle point tip that penetrates the bone for easier drilling. It helps avoid skipping over the bone especially if drilling at an angle other than 90°. Made from hard 420 stainless steel.
Catalog No. 50-005-81844