Learn More
Uline Anti-static Pick and Pack Foam Sheets 24 x 24 x 1"""", 12/case
Supplier:  Uline S15323CS
Encompass
Anti-static polyurethane open-cell foam sheet dissipates static charges, provides lightweight cushioning and ESD protection, perforated sheets
Catalog No. NC2382865