Learn More
Medchemexpress LLC Antithrombin III (Serpin C1) recombinant protein | 90170-80-2 | >95.0% | 10 UG

Supplier: Medchemexpress LLC HYP7558110UG
Recombinant antithrombin III (Serpin C1) derived from cynomolgus and expressed in HEK293 cells. Supplied as a 10 μg research pack, the C-terminal His-tagged protein is suitable for biochemical and cell-based assays involving anticoagulant serpin activity.
- High purity (>95.0% by reducing SDS-PAGE).
- Expressed in HEK293 mammalian cells.
- C-terminal His tag for detection or purification.
- Species: cynomolgus.
- Approximate molecular weight 55-75 kDa.
- Store at -20°C; aliquot to avoid freeze-thaw.
- Certificate of analysis and SDS available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.