Learn More
Medchemexpress LLC Antitubercular agent-30 | 384857-54-9 | C12H10N2O3S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15123610MM/1ML
Encompass_Preferred
Antitubercular agent-30 is an antibacterial agent effective against Mycobacterium tuberculosis with an MIC of 50 μg/mL. It exhibits minimal cytotoxic effect on murine macrophage cells, with an LD85 of approximately 100 μg/mL, and showed no cytotoxic effect on murine macrophages below 10 μg/mL when tested at concentrations between 0.1-100 μg/mL for 18 hours.
  • Antibacterial agent
  • Effective against Mycobacterium tuberculosis
  • Minimal cytotoxic effect on murine macrophage cells
  • MIC of 50 μg/mL
  • LD85 of approximately 100 μg/mL
Catalog No. 50-003-75871