Learn More
TARGETMOL CHEMICALS INC ANTITUMOR AGENT-19 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T103415MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Antitumor agent-19 is a modulator of tumor-associated macrophages with EC50s of 17.18 uM and 18.87 uM in the RAW 264.7 cells and the BMDM cells. purity: 98%
Catalog No. 50-258-1733