Learn More
Creative Biomart Agkistrodon piscivorus antivenin
Supplier:  Creative Biomart THP0157
Encompass_Preferred
The product is derived and purified immunoglobulin fragments obtained from other domestic animals such as sheep previously immunized with Agkistrodon piscivorus. The final purified antivenin product is obtained by mixing other different monospecific snake antivenins and isolating the antivenin of interest through fractionation and chromatography techniques.
Catalog No. 50-208-4208