Learn More
Medchemexpress LLC Antiviral agent 24 | 23661-03-2 | C18H18F3N5O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15192610MM/1ML
Encompass_Preferred
Antiviral agent 24 is a potent antiviral agent that inhibits METTL3/METTL14 activity in a dose-dependent manner. It exhibits EC50 values of 0.101, 19.9, and 91.2 μM for EV71, CVA21, and EV68, respectively. This product is for research use only.
  • Potent antiviral agent
  • Inhibits METTL3/METTL14 activity
  • Effective against EV71, CVA21, EV68
Catalog No. 50-003-73295