Learn More
Medchemexpress LLC Antiviral agent 24 | 23661-03-2 | C18H18F3N5O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151926
Encompass_Preferred
Antiviral agent 24 is a potent antiviral agent with EC50 values of 0.101, 19.9, and 91.2 μM for EV71, CVA21, and EV68, respectively. It inhibits METTL3/METTL14 activity in a dose-dependent manner. This product is for research use only.
  • Potent antiviral agent
  • Inhibits METTL3/METTL14 activity in a dose-dependent manner
  • Purity: 99.63%
Catalog No. 50-003-75990