Learn More
Sigma Aldrich Fine Chemicals Biosciences Antonia RedTM Dextran 1505MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 805435MG
Encompass
Antonia Red(TM) dextrans are purple powders that are readily soluble in water or electrolyteSOLUTIONons. Antonia Red dextran are insoluble in most organic solvents.
Catalog No. 50-290-1633