Learn More
AdipoGen ANTS
SDP
Supplier:  AdipoGen CDXA0071G010
Encompass_Preferred
Chemical. CAS 5398-34-5. Formula C10H7NNa2O9S3. Molecular Weight 427.34. ANTS is a highly negatively charged dye with an amino group that can be coupled to an aldehyde or ketone group to form an unstable Schiff base. The Schiff base is usually chemically reduced by sodium borohydride NaBH4 or sodium cyanoborohydride NaBCNH3 to form a stable linkage. This labeling technique has been widely used for the labeling and subsequent sequencing of oligosaccharides and glycoproteins. The negative charges of the dye facilitate the electrophoretic separation of the degradation products of carbohydrate polymers. The polyanionic dye ANTS is used together with the fluorescent cationic quencher DPX for membrane fusion studies or permeability assays. The mixture of ANTS-DPX is minimally fluorescent initially, but becomes increasingly more fluorescent upon dilution i.e. membrane fusion or leakage. ANTS also is used as a neuronal tracer.
Catalog No. 50-256-1841