Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ANTS | 5398-34-5 | 99.1% | 100 MG
SDP

Supplier:  Medchemexpress LLC HYD0789100MG

Encompass_Preferred

ANTS is a fluorescent dye utilized for measuring membrane leakage. It has an excitation wavelength of 380 nm and an emission wavelength of 520 nm. When encapsulated in liposomes with DPX, ANTS provides an effective method for assessing membrane permeability.

  • Solid appearance, white to off-white color.
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO at 20 mg/mL (46.80 mM) with ultrasonic, warming, and heating to 80°C.

Catalog No. 50-004-10888


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.