Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ANTS | 5398-34-5 | 99.1% | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYD0789

Encompass_Preferred

ANTS is a fluorescent dye with an excitation wavelength of 380 nm and an emission wavelength of 520 nm. When encapsulated with DPX in liposomes, ANTS can be used as an effective method for measuring membrane leakage.

  • Fluorescent dye for cell staining analysis
  • Used as a membrane dye
  • Excitation wavelength: 380 nm
  • Emission wavelength: 520 nm
  • Can measure membrane leakage when encapsulated with DPX in liposomes

Catalog No. 50-004-13558


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.