Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ANTS | 5398-34-5 | 99.1% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYD078950MG

Encompass_Preferred

ANTS is a fluorescent dye (Excitation wavelength: 380 nm; Emission wavelength: 520 nm). When encapsulated in liposomes with DPX, ANTS can be an effective approach for measuring membrane leakage. It is suitable for high-speed applications in laboratory centrifugation and storage.

  • Fluorescent dye for cell staining analysis
  • Excitation wavelength of 350 nm
  • Emission wavelength of 515 nm
  • White to off-white solid appearance
  • Purity of 99.09%
  • Can be used for membrane leakage measurements

Catalog No. 50-004-10861


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.